siRNA Details
Gene Symbol: Dmrtc1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014222.2 | NP_001014244.2 | 9 | 1146 | ||
GenBank mRNA | BC079417.1 | - | 9 | 1132 | 7113562 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dmrtc1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014222.2 | NP_001014244.2 | 9 | 1146 | ||
GenBank mRNA | BC079417.1 | - | 9 | 1132 | 7113562 |