siRNA Details
| Gene Symbol: Dnase1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014223.2 | NP_001014245.2 | 5, 6 | 689 | ||
| XM_006229602.1 | XP_006229664.1 | 4, 5 | 666 | |||
| GenBank mRNA | BC079074.1 | - | 5, 6 | 645 | 7122158 | |
| DQ116785.1 | - | 5, 6 | 689 | |||
| FQ232580.1 | - | 4, 5 | 615 | |||

Dnase1l1
Genomic Map