siRNA Details
| Gene Symbol: Cluap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014225.1 | NP_001014247.1 | 12 | 1774 | ||
| XM_006245832.2 | XP_006245894.1 | 12 | 1537 | |||
| XM_008767512.1 | XP_008765734.1 | 12 | 1470 | |||
| XM_008767513.1 | XP_008765735.1 | 12 | 1832 | |||
| XM_008767514.1 | XP_008765736.1 | 11 | 1374 | |||
| XM_008767515.1 | XP_008765737.1 | 11 | 1378 | |||
| XM_008767516.1 | XP_008765738.1 | 11 | 1376 | |||
| XM_008767517.1 | XP_008765739.1 | 10 | 1239 | |||
| XM_008767518.1 | XP_008765740.1 | 10 | 1296 | |||
| GenBank mRNA | BC079022.1 | - | 12 | 1774 | 7113868 | |

Cluap1
Genomic Map