siRNA Details
| Gene Symbol: ODAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017855.3 | NP_060325.3 | 8 | 614 | ||
| XM_005265698.3 | XP_005265755.1 | 9 | 678 | |||
| XM_006714245.3 | XP_006714308.1 | 8 | 607 | |||
| XM_017008331.1 | XP_016863820.1 | 7 | 570 | |||
| GenBank mRNA | AK000520.1 | - | 5 | 265 | ||
| AK225547.1 | - | 5 | 245 | |||
| BC017796.1 | - | 4 | 221 | 4689080 | ||
| BG194365.1 | - | 8 | 617 | |||
| EF113908.1 | - | 8 | 616 | |||

ODAM
Genomic Map