siRNA Details
| Gene Symbol: C1orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017860.3 | NP_060330.2 | 1 | 268 | ||
| GenBank mRNA | AK000526.1 | - | 1 | 277 | ||
| AK301712.1 | - | 1 | 340 | |||
| AK315680.1 | - | 1 | 280 | |||
| AY358436.1 | - | 1 | 311 | |||
| BC002469.2 | - | 1 | 268 | 3347975 | ||

C1orf56
Genomic Map