siRNA Details
| Gene Symbol: Lzts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014247.2 | NP_001014269.2 | 4 | 1826 | ||
| XM_006231547.2 | XP_006231609.1 | 5 | 2876 | |||
| XM_006231548.2 | XP_006231610.1 | 6 | 2077 | |||
| XM_006231549.2 | XP_006231611.1 | 5 | 3305 | |||
| XM_008760444.1 | XP_008758666.1 | 6 | 2068 | |||
| GenBank mRNA | AY928801.1 | - | 4 | 1784 | ||
| BC083580.1 | - | 7 | 1632 | 7129196 | ||
| DN932851.1 | - | 2 | 502 | |||
| DQ176638.2 | - | 4 | 1784 | |||

Lzts2
Genomic Map