siRNA Details
| Gene Symbol: LOC365791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761013.1 | XP_008759235.1 | 15 | 2143 | ||
| GenBank mRNA | BC081933.1 | - | 2014 | 7114567 | ||
| Gene Symbol: LOC102550446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761011.1 | XP_008759233.1 | 9 | 1675 | ||

LOC365791
Genomic Map