siRNA Details
| Gene Symbol: Selt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014253.2 | NP_001014275.2 | 4, 5 | 523 | ||
| GenBank mRNA | AY995234.1 | - | 4, 5 | 523 | ||
| BC086953.1 | - | 4, 5 | 417 | 7321890 | ||
| FQ211813.1 | - | 541 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Selt| Gene Symbol: Selt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014253.2 | NP_001014275.2 | 4, 5 | 523 | ||
| GenBank mRNA | AY995234.1 | - | 4, 5 | 523 | ||
| BC086953.1 | - | 4, 5 | 417 | 7321890 | ||
| FQ211813.1 | - | 541 | ||||