siRNA Details
| Gene Symbol: Trub2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014257.2 | NP_001014279.1 | 1, 2 | 146 | ||
| GenBank mRNA | BC084712.1 | - | 1, 2 | 121 | 7303031 | |
| CB613146.1 | - | 1, 2 | 146 | |||
| FQ231592.1 | - | 1, 2 | 160 | |||

Trub2
Genomic Map