siRNA Details
| Gene Symbol: Cep83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014266.1 | NP_001014288.1 | 8, 9 | 1243 | ||
| XM_008765317.1 | XP_008763539.1 | 8, 9 | 1239 | |||
| XM_008765318.1 | XP_008763540.1 | 7, 8 | 1085 | |||
| GenBank mRNA | BC081969.1 | - | 8, 9 | 1243 | 7125504 | |

Cep83
Genomic Map