siRNA Details
| Gene Symbol: ASB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202403.1 | NP_001189332.1 | 5 | 1398 | ||
| NM_017873.3 | NP_060343.1 | 6 | 1485 | |||
| NM_177999.2 | NP_821066.1 | 5 | 1376 | |||
| GenBank mRNA | AK000555.1 | - | 6 | 1482 | ||
| AK023004.1 | - | 6 | 1431 | |||
| AK225618.1 | - | 5 | 1373 | |||
| AK292807.1 | - | 6 | 1482 | |||
| AK299193.1 | - | 5 | 1280 | |||
| BC001719.1 | - | 5 | 1331 | |||
| BC065913.1 | - | 6 | 1388 | 5211288 | ||
| BX648923.1 | - | 5 | 2006 | |||

ASB6
Genomic Map