siRNA Details
| Gene Symbol: Slco6d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014270.1 | NP_001014292.1 | 900 | |||
| XM_006245580.2 | XP_006245642.1 | 1138 | ||||
| XM_006245583.2 | XP_006245645.1 | 877 | ||||
| GenBank mRNA | BC083829.1 | - | 900 | 7112354 | ||

Slco6d1
Genomic Map