siRNA Details
| Gene Symbol: FLG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014342.2 | NP_001014364.1 | 3 | 8209 | ||
| XM_011509531.2 | XP_011507833.1 | 4 | 7775 | |||
| GenBank mRNA | AY827490.1 | - | 3 | 8210 | ||
| BC108269.1 | - | 1 | 660 | |||

FLG2
Genomic Map