siRNA Details
| Gene Symbol: FAM99A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026643.1 | 3 | 587 | |||
| GenBank mRNA | AK124823.1 | - | 1 | 1191 | ||
| CR627417.1 | - | 3 | 614 | |||
| Gene Symbol: FAM99B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026642.1 | 3 | 613 | |||

FAM99A
Genomic Map