siRNA Details
| Gene Symbol: Kpna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014792.1 | NP_001014792.1 | 15 | 1273 | ||
| XM_006252127.2 | XP_006252189.1 | 15 | 1367 | |||
| XM_008770761.1 | XP_008768983.1 | 15 | 1222 | |||
| GenBank mRNA | AY779027.1 | - | 15 | 1273 | ||

Kpna3
Genomic Map