siRNA Details
| Gene Symbol: Kpna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014793.1 | NP_001014793.1 | 5 | 256 | ||
| GenBank mRNA | AY779028.1 | - | 5 | 256 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008467, AK147920, AF020771, BC052162, BC026821, and others.

Kpna4
Genomic Map