siRNA Details
| Gene Symbol: Ubl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015030.1 | NP_001015030.1 | 2 | 849 | ||
| GenBank mRNA | BC091342.1 | - | 2 | 849 | 7315459 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubl3| Gene Symbol: Ubl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015030.1 | NP_001015030.1 | 2 | 849 | ||
| GenBank mRNA | BC091342.1 | - | 2 | 849 | 7315459 | |