siRNA Details
| Gene Symbol: UFSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015072.3 | NP_001015072.2 | 1 | 494 | ||
| GenBank mRNA | AK292467.1 | - | 1 | 494 | ||
| BC017456.2 | - | 1 | 429 | |||
| BC067904.1 | - | 1 | 457 | 5310615 | ||
| DB073745.1 | - | 1 | 494 | |||

UFSP1
Genomic Map