siRNA Details
| Gene Symbol: Fam71d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017469.1 | NP_001017469.1 | 6 | 1095 | ||
| GenBank mRNA | BC078980.1 | - | 6 | 1095 | 7112035 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam71d| Gene Symbol: Fam71d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017469.1 | NP_001017469.1 | 6 | 1095 | ||
| GenBank mRNA | BC078980.1 | - | 6 | 1095 | 7112035 | |