siRNA Details
| Gene Symbol: Cep70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017470.1 | NP_001017470.1 | 3 | 229 | ||
| XM_006243635.1 | XP_006243697.1 | 3 | 295 | |||
| XM_006243639.1 | XP_006243701.1 | 3 | 295 | |||
| XM_006243641.1 | XP_006243703.1 | 3 | 295 | |||
| XM_008766552.1 | XP_008764774.1 | 3 | 295 | |||
| XM_008766553.1 | XP_008764775.1 | 3 | 280 | |||
| XM_008766554.1 | XP_008764776.1 | 4 | 412 | |||
| XM_008766555.1 | XP_008764777.1 | 3 | 295 | |||
| GenBank mRNA | BC087075.1 | - | 3 | 229 | 7134655 | |

Cep70
Genomic Map