siRNA Details
| Gene Symbol: Cep70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017470.1 | NP_001017470.1 | 4, 5 | 327 | ||
| XM_006243635.1 | XP_006243697.1 | 4, 5 | 393 | |||
| XM_006243639.1 | XP_006243701.1 | 4, 5 | 393 | |||
| XM_006243640.2 | XP_006243702.1 | 2, 3 | 168 | |||
| XM_008766552.1 | XP_008764774.1 | 5, 6 | 435 | |||
| XM_008766553.1 | XP_008764775.1 | 5, 6 | 420 | |||
| XM_008766554.1 | XP_008764776.1 | 6, 7 | 552 | |||
| XM_008766555.1 | XP_008764777.1 | 5, 6 | 435 | |||
| GenBank mRNA | BC087075.1 | - | 4, 5 | 327 | 7134655 | |

Cep70
Genomic Map