siRNA Details
| Gene Symbol: ZNF749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001023561.3 | NP_001018855.2 | 3 | 1681 | ||
| NM_001321952.1 | NP_001308881.1 | 3 | 1755 | |||
| NM_001321953.1 | NP_001308882.1 | 2 | 1628 | |||
| NM_001321954.1 | NP_001308883.1 | 2 | 1554 | |||
| XM_011526956.2 | XP_011525258.1 | 3 | 1430 | |||
| XM_017026802.1 | XP_016882291.1 | 4 | 2681 | |||
| GenBank mRNA | AK122740.1 | - | 1 | 423 | ||
| AK122794.1 | - | 3 | 1677 | |||

ZNF749
Genomic Map