siRNA Details
| Gene Symbol: POT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090675.1 | - | 6 | 1154 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

POT1-AS1| Gene Symbol: POT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090675.1 | - | 6 | 1154 | ||