siRNA Details
| Gene Symbol: TTC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271420.1 | NP_001258349.1 | 5, 6 | 1136 | ||
| NM_017775.3 | NP_060245.3 | 5, 6 | 999 | |||
| XM_017024801.1 | XP_016880290.1 | 5, 6 | 912 | |||
| XM_017024802.1 | XP_016880291.1 | 5, 6 | 912 | |||
| GenBank mRNA | AK000350.1 | - | 3, 4 | 215 | ||
| AK055780.1 | - | 5, 6 | 692 | |||
| AK056878.1 | - | 5, 6 | 911 | |||
| AK297783.1 | - | 5, 6 | 555 | |||
| BC011698.2 | - | 5, 6 | 538 | |||
| BC073796.1 | - | 5, 6 | 542 | |||
| BC105128.1 | - | 5, 6 | 999 | 8144131 | ||
| BC112107.1 | - | 5, 6 | 999 | 8327575 | ||

TTC19
Genomic Map