siRNA Details
| Gene Symbol: Rassf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024275.1 | NP_001019446.1 | 11 | 1175 | ||
| GenBank mRNA | BC093620.1 | - | 11 | 1175 | 7315142 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rassf4| Gene Symbol: Rassf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024275.1 | NP_001019446.1 | 11 | 1175 | ||
| GenBank mRNA | BC093620.1 | - | 11 | 1175 | 7315142 | |