siRNA Details
| Gene Symbol: Gcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024277.1 | NP_001019448.1 | 7 | 947 | ||
| GenBank mRNA | BC092591.1 | - | 7 | 947 | 7372269 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gcat| Gene Symbol: Gcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024277.1 | NP_001019448.1 | 7 | 947 | ||
| GenBank mRNA | BC092591.1 | - | 7 | 947 | 7372269 | |