siRNA Details
| Gene Symbol: SBK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024401.2 | NP_001019572.1 | 4 | 4162 | ||
| XM_005255315.3 | XP_005255372.1 | 4 | 4196 | |||
| GenBank mRNA | AL390134.1 | - | 1 | 1301 | ||
| AY874862.2 | - | 4 | 4162 | |||

SBK1
Genomic Map