siRNA Details
| Gene Symbol: C1orf53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024594.2 | NP_001019765.1 | 1 | 270 | ||
| GenBank mRNA | BC038214.1 | - | 1 | 169 | ||
| BC130608.1 | - | 1 | 270 | 40146639 | ||
| BC130634.1 | - | 1 | 270 | 40146665 | ||

C1orf53
Genomic Map