siRNA Details
| Gene Symbol: FEZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024613.3 | NP_001019784.2 | 1 | 531 | ||
| XM_005250337.3 | XP_005250394.1 | 2 | 2202 | |||
| GenBank mRNA | AK300742.1 | - | 1 | 660 | ||
| AY726588.1 | - | 1 | 452 | |||
| BC127714.1 | - | 1 | 519 | 40127896 | ||
| BC136684.1 | - | 1 | 587 | 9020674 | ||
| BC144367.1 | - | 1 | 587 | 9052888 | ||

FEZF1
Genomic Map