siRNA Details
| Gene Symbol: OSBPL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174060.1 | NP_001167531.1 | 11 | 3074 | ||
| NM_017784.4 | NP_060254.2 | 12 | 3266 | |||
| XM_005264845.1 | XP_005264902.1 | 7 | 1950 | |||
| GenBank mRNA | AB209872.1 | - | 8 | 3863 | ||
| AF392451.1 | - | 13 | 2976 | |||
| AK000370.1 | - | 10 | 2142 | |||
| AK304067.1 | - | 11 | 2527 | |||
| AL389943.1 | - | 1 | 1646 | |||
| AL833247.1 | - | 12 | 2369 | |||
| BC003168.1 | - | 12 | 2670 | 2905981 | ||

OSBPL10
Genomic Map