siRNA Details
| Gene Symbol: Tmem165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024802.1 | NP_001019973.1 | 6 | 1773 | ||
| GenBank mRNA | BC097478.1 | - | 6 | 1773 | 7458161 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem165| Gene Symbol: Tmem165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024802.1 | NP_001019973.1 | 6 | 1773 | ||
| GenBank mRNA | BC097478.1 | - | 6 | 1773 | 7458161 | |