siRNA Details
| Gene Symbol: Ehd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024897.1 | NP_001020068.1 | 6 | 1456 | ||
| GenBank mRNA | AF081251.1 | - | 1 | 44 | ||
| BC097385.1 | - | 6 | 1456 | 7446444 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ehd2| Gene Symbol: Ehd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024897.1 | NP_001020068.1 | 6 | 1456 | ||
| GenBank mRNA | AF081251.1 | - | 1 | 44 | ||
| BC097385.1 | - | 6 | 1456 | 7446444 | ||