siRNA Details
| Gene Symbol: FLVCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195283.1 | NP_001182212.1 | 10 | 2539 | ||
| NM_017791.2 | NP_060261.2 | 10 | 3421 | |||
| GenBank mRNA | AK000378.1 | - | 10 | 3369 | ||
| AY260577.1 | - | 10 | 3401 | |||
| CR739521.1 | - | 1 | 367 | |||

FLVCR2
Genomic Map