siRNA Details
Gene Symbol: Rc3h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024952.2 | NP_001020123.1 | 9 | 1745 | ||
GenBank mRNA | AK139648.1 | - | 9 | 1717 | ||
AK173335.1 | - | 9 | 1691 | |||
AY948287.1 | - | 9 | 1686 | |||
BC138663.1 | - | 8 | 1469 | 8861684 |

More Information
Other Species Targeted

Gene Ontology Categories:
