siRNA Details
| Gene Symbol: Fam26f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024976.1 | NP_001020147.1 | 2 | 887 | ||
| XM_008772880.1 | XP_008771102.1 | 2 | 776 | |||
| GenBank mRNA | BC093379.1 | - | 2 | 887 | 7388112 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam26f| Gene Symbol: Fam26f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024976.1 | NP_001020147.1 | 2 | 887 | ||
| XM_008772880.1 | XP_008771102.1 | 2 | 776 | |||
| GenBank mRNA | BC093379.1 | - | 2 | 887 | 7388112 | |