siRNA Details
| Gene Symbol: Them4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025017.1 | NP_001020188.1 | 6 | 1426 | ||
| GenBank mRNA | BC093385.1 | - | 6 | 1426 | 7389383 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Them4| Gene Symbol: Them4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025017.1 | NP_001020188.1 | 6 | 1426 | ||
| GenBank mRNA | BC093385.1 | - | 6 | 1426 | 7389383 | |