siRNA Details
| Gene Symbol: Ntmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025019.1 | NP_001020190.1 | 4 | 1221 | ||
| XM_006233919.2 | XP_006233981.1 | 5 | 2046 | |||
| XM_008761670.1 | XP_008759892.1 | 3 | 1111 | |||
| GenBank mRNA | BC091294.1 | - | 4 | 1221 | 7307721 | |
| FQ216821.1 | - | 4 | 1236 | |||

Ntmt1
Genomic Map