siRNA Details
| Gene Symbol: Stap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025026.1 | NP_001020197.1 | 7 | 735 | ||
| GenBank mRNA | BC091228.1 | - | 7 | 735 | 7321609 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stap2| Gene Symbol: Stap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025026.1 | NP_001020197.1 | 7 | 735 | ||
| GenBank mRNA | BC091228.1 | - | 7 | 735 | 7321609 | |