siRNA Details
| Gene Symbol: Sult2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025131.1 | NP_001020302.1 | 3 | 464 | ||
| XM_008758949.1 | XP_008757171.1 | 3 | 488 | |||
| GenBank mRNA | D14989.1 | - | 3 | 464 | ||
| FQ209413.1 | - | 3 | 477 | |||
| FQ218086.1 | - | 3 | 451 | |||
| FQ219053.1 | - | 3 | 477 | |||
| FQ219201.1 | - | 3 | 477 | |||
| FQ219357.1 | - | 3 | 450 | |||
| FQ219375.1 | - | 3 | 488 | |||
| FQ219766.1 | - | 3 | 476 | |||

Sult2a2
Genomic Map