siRNA Details
| Gene Symbol: Sult2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025131.1 | NP_001020302.1 | 5 | 726 | ||
| GenBank mRNA | D14989.1 | - | 5 | 726 | ||
| FQ209413.1 | - | 5 | 739 | |||
| FQ218086.1 | - | 5 | 713 | |||
| FQ219053.1 | - | 5 | 739 | |||
| FQ219201.1 | - | 5 | 739 | |||
| FQ219357.1 | - | 5 | 712 | |||
| FQ219375.1 | - | 5 | 750 | |||
| FQ219766.1 | - | 5 | 738 | |||

Sult2a2
Genomic Map