siRNA Details
| Gene Symbol: Sult2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025131.1 | NP_001020302.1 | 2 | 239 | ||
| XM_008758949.1 | XP_008757171.1 | 2 | 263 | |||
| GenBank mRNA | D14989.1 | - | 2 | 239 | ||
| FQ209413.1 | - | 2 | 252 | |||
| FQ218086.1 | - | 2 | 226 | |||
| FQ219053.1 | - | 2 | 252 | |||
| FQ219201.1 | - | 2 | 252 | |||
| FQ219357.1 | - | 2 | 225 | |||
| FQ219375.1 | - | 2 | 263 | |||
| FQ219766.1 | - | 2 | 251 | |||

Sult2a2
Genomic Map