siRNA Details
| Gene Symbol: Cnih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025132.1 | NP_001020303.1 | 6 | 1216 | ||
| XM_008760153.1 | XP_008758375.1 | 5 | 1794 | |||
| XM_008760154.1 | XP_008758376.1 | 6 | 1854 | |||
| GenBank mRNA | BC091325.1 | - | 6 | 1216 | 7313787 | |

Cnih2
Genomic Map