siRNA Details
| Gene Symbol: Klk1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008693.2 | NP_032719.1 | 2, 3 | 259 | ||
| GenBank mRNA | BC132657.1 | - | 2, 3 | 259 | 40130934 | |
| BC132659.1 | - | 2, 3 | 259 | 40130936 | ||
| X01389.1 | - | 2, 3 | 220 | |||

Klk1b3
Genomic Map