siRNA Details
| Gene Symbol: Chic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009767.2 | NP_033897.1 | 2 | 437 | ||
| GenBank mRNA | AK034427.1 | - | 2 | 410 | ||
| AK135974.1 | - | 2 | 389 | |||
| BC132322.1 | - | 2 | 356 | 40130599 | ||
| BC138730.1 | - | 2 | 356 | 8861752 | ||

Chic1
Genomic Map