siRNA Details
| Gene Symbol: RASIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017805.2 | NP_060275.2 | 12 | 3280 | ||
| XM_011527053.2 | XP_011525355.2 | 12 | 3430 | |||
| XM_011527055.2 | XP_011525357.2 | 9 | 3136 | |||
| XM_017026914.1 | XP_016882403.1 | 12 | 3400 | |||
| XM_017026915.1 | XP_016882404.1 | 9 | 3106 | |||
| GenBank mRNA | AK000408.1 | - | 8 | 1395 | ||
| AK123179.1 | - | 4 | 1577 | |||
| AY378097.1 | - | 12 | 3145 | |||
| BC021860.1 | - | 8 | 1410 | |||
| BC028614.1 | - | 12 | 3152 | 4815880 | ||

RASIP1
Genomic Map