siRNA Details
| Gene Symbol: SLC8A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015063.2 | NP_055878.1 | 4 | 1705 | ||
| XM_005259172.1 | XP_005259229.1 | 4 | 1514 | |||
| GenBank mRNA | AB029010.1 | - | 4 | 1520 | ||
| AK293457.1 | - | 4 | 446 | |||
| AK293860.1 | - | 3 | 838 | |||
| AK302552.1 | - | 4 | 1705 | |||
| AL512764.1 | - | 3 | 815 | |||

SLC8A2
Genomic Map