siRNA Details
| Gene Symbol: ROPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017578.3 | NP_060048.2 | 3 | 287 | ||
| NR_133918.1 | 3 | 364 | ||||
| NR_133919.1 | 2 | 318 | ||||
| XM_011512934.2 | XP_011511236.1 | 3 | 362 | |||
| GenBank mRNA | AF303889.2 | - | 3 | 287 | ||
| AK310283.1 | - | 2 | 257 | |||
| BC132744.1 | - | 1 | 43 | 40146766 | ||
| BC132746.1 | - | 1 | 44 | 40146768 | ||
| DB042884.1 | - | 3 | 335 | |||

ROPN1
Genomic Map