siRNA Details
| Gene Symbol: ZNF708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128184.1 | - | 5 | 3487 | ||
| AK131527.1 | - | 4 | 3661 | |||
| BC039403.1 | - | 1 | 284 | |||
| BX103920.1 | - | 1 | 358 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF708| Gene Symbol: ZNF708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK128184.1 | - | 5 | 3487 | ||
| AK131527.1 | - | 4 | 3661 | |||
| BC039403.1 | - | 1 | 284 | |||
| BX103920.1 | - | 1 | 358 | |||