siRNA Details
| Gene Symbol: Rnase4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021472.4 | NP_067447.2 | 2 | 582 | ||
| NM_201239.3 | NP_957691.1 | 2 | 879 | |||
| GenBank mRNA | AB041044.1 | - | 2 | 488 | ||
| AK050451.1 | - | 2 | 479 | |||
| AK076504.1 | - | 2 | 552 | |||
| AY762362.1 | - | 2 | 481 | |||
| BC005569.1 | - | 2 | 456 | 3967265 | ||
| BC061105.1 | - | 2 | 463 | 30296907 | ||
| BC066843.1 | - | 2 | 543 | 30059308 | ||

Rnase4
Genomic Map