siRNA Details
| Gene Symbol: Tas2r140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021562.1 | NP_067537.1 | 1 | 889 | ||
| GenBank mRNA | BC145965.1 | - | 1 | 912 | 40131268 | |
| BC145967.1 | - | 1 | 912 | 40131270 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r140| Gene Symbol: Tas2r140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021562.1 | NP_067537.1 | 1 | 889 | ||
| GenBank mRNA | BC145965.1 | - | 1 | 912 | 40131268 | |
| BC145967.1 | - | 1 | 912 | 40131270 | ||